Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoserine Antibody, Novus Biologicals™
SDP

Catalog No. NB1001953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
0.4 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001953 0.4 mL
NB001692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001953 Supplier Novus Biologicals Supplier No. NB1001953
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Phosphoserine Polyclonal specifically detects Phosphoserine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Phosphoserine
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:250, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500
Formulation PBS, 50% Glycerol with 0.01 mg Sodium Azide
Gene Alias Phosphoserine; Phosphothreonine
Host Species Rabbit
Immunogen Phosphoserine conjugated to KLH, and phosvitin mixture
Purification Method Affinity Purified
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes proteins phosphorylated on serine residues. Does not cross-react with phosphotyrosine.
Target Species Non-species specific, Mouse, Non-species specific
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.