Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoserine Mouse, Clone: PSR-45, Abnova™
Mouse monoclonal antibody raised against phosphoserine.
Supplier: Abnova Corporation MAB12456
Specifications
Phosphoserine | |
Monoclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Lyophilized |
Western Blot | |
PSR-45 | |
Mouse monoclonal antibody raised against phosphoserine. | |
Lyophilized from ascites fluid (1.2% sodium acetate, 2mg BSA, 0.01mg sodium azide) | |
Phosphoserine (conjugated with KLH). | |
RUO | |
Store at −20°C. After reconstitution with 1mLof 1.2% sodium acetate or neutral PBS and concentration will be 100μg/mL, store at −20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction