Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphoserine phosphatase Antibody (3G12), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104297V
Description
Phosphoserine phosphatase Monoclonal specifically detects Phosphoserine phosphatase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Phosphoserine phosphatase | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSPH | |
Recombinant human PSP (1-225aa) purified from E. coli | |
0.1 mL | |
Lipid and Metabolism | |
5723 | |
Human | |
Purified |
Western Blot, ELISA | |
3G12 | |
Western Blot, ELISA | |
L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human PSP mAb, clone 3G12, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSP protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction