Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphothreonine Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11096896100UL
Description
Phosphothreonine Polyclonal specifically detects Phosphothreonine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Phosphothreonine | |
Polyclonal | |
Unconjugated | |
PBS, 50% Glycerol | |
Phosphothreonine conjugated to KLH | |
100 μL | |
Non-species specific | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
Western Blot 2 μg/mL, ELISA 0.5 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 10 μg/mg, Immunohistochemistry-Paraffin | |
Rabbit | |
Affinity Purified | |
Recognizes proteins phosphorylated on threonine residues. Does not cross-react with phosphotyrosine. | |
Aliquot and store at -20°C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction