Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phosphothreonine Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178035
Description
Phosphothreonine Polyclonal specifically detects Phosphothreonine in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Phosphothreonine | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative | |
Phosphothreonine antibody was prepared from whole rabbit serum produced by repeated immunizations with phosphothreonine conjugated KLH. | |
0.1 mg | |
Amino Acids Drugs and other small molecules, Signal Transduction | |
Reactive in phosphothreonine. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 2.0 ug/ml, ELISA 0.5 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 10.0 ug/mg, Immunohistochemistry-Paraffin 1:10-1:500 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Non-species specific | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction