Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phosphothreonine Monoclonal Antibody (18F6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA191728
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphothreonine is a phosphoamino acid, the phosphorylated ester of threonine. There are three amino acids that are typically phosphorylated in eukaryotes: serine, threonine, and tyrosine. Threonine residues in endogenous proteins undergo phosphorylation through the action of a threonine kinase. Small amounts of free phosphothreonine can be detected in urine Antibodies against phosphothreonine can be used as an identifier in the analysis of peptides, proteins, and enzymes. [National Center for Biotechnology Information.
Specifications
Phosphothreonine | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.01% sodium azide | |
Mouse | |
50 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA, Western Blot | |
18F6 | |
Unconjugated | |
C4H10NO6P; O-phospho-L-threonine; Phospho-threonine; pThr; Threonine | |
Phosphothreonine conjugated KLH. | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction