Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphothreonine Recombinant Rabbit Monoclonal Antibody (RM102)
GREENER_CHOICE

Catalog No. PIMA527976 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527976 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527976 Supplier Invitrogen™ Supplier No. MA527976
  • $679.00
    $514.65 / Each of 1

    Save $164.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Phosphothreonine is a phosphoamino acid, the phosphorylated ester of threonine. There are three amino acids that are typically phosphorylated in eukaryotes: serine, threonine, and tyrosine. Threonine residues in endogenous proteins undergo phosphorylation through the action of a threonine kinase. Small amounts of free phosphothreonine can be detected in urine Antibodies against phosphothreonine can be used as an identifier in the analysis of peptides, proteins, and enzymes. [National Center for Biotechnology Information.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphothreonine
Applications ChIP Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Luminex, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RM102
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene Alias C4H10NO6P; O-phospho-L-threonine; Phospho-threonine; pThr; Threonine
Host Species Rabbit
Immunogen Mixture of phosphothreonine-BSA conjugate and a phosphothreonine containing peptide.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.