Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphotyrosine Monoclonal Antibody (pY20)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 037799

Encompass_Preferred

 View more versions of this product

Catalog No. 03-779-9


Only null left
Edge
Add to Cart

Description

Description

This antibody detects proteins which contain phosphorylated tyrosine residues. This antibody is known to show cross-reactivity towards phosphothreonine.

The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phosphotyrosine
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide; pH 7.4
Mouse
Protein A
RUO
Chemical
Antibody
IgG2b
ELISA, Immunoprecipitation, Western Blot
pY20
Unconjugated
Phosphotyrosine; pTyr; pY
Keyhole limpet hemocyanin coupled with equal amounts of phosphotyrosine, glycine, and alanine.
1 mg
Primary
-20°C
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.