Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phosphotyrosine Monoclonal Antibody (pY20)
GREENER_CHOICE

Catalog No. MA182787
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA182787 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA182787 Supplier Invitrogen™ Supplier No. MA182787
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

For the detection of phosphoproteins, tyrosine phosphatase inhibitors such as 1mM sodium orthovanadate should be added to the sample buffer. Milk or other casein-based blocking solutions are not recommended as casein is a phosphoprotein and its use can result in high background. Mouse anti Phosphotyrosine antibody, clone PY20 recognizes phosphotyrosine, enabling the detection, characterisation and isolation of proteins containing phosphorylated tyrosine residues.

The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone pY20
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation BBS with <0.1% sodium azide; pH 8.0
Gene Alias Phosphotyrosine; pTyr; pY
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Target Molecule Chemical
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.