Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Mouse, Clone: 18E10D2, Abnova™

Catalog No. 89035664 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-664 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-664 Supplier Abnova Corporation Supplier No. MAB14228

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against Phosphotyrosine.

  • High specificity
  • High reactivity

Specifications

Antigen Phosphotyrosine
Applications Western Blot
Classification Monoclonal
Clone 18E10D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Phosphotyrosine.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 0.5 mg/mL PBS, pH 7.4 (0.02% sodium azide).
Host Species Mouse
Immunogen Phosphotyrosine containing carrier protein.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody is specific for proteins containing phosphotyrosine residues. It does not cross-react with phosphoserine, phosphothreonine or nonphosphorylated tyrosine residues.
Content And Storage Store at -20°C or lower.
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL.
The reconstituted antibody can be stored for 2-3 weeks at 4°C. For long term storage, aliquot and st
Form Lyophilized
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.