Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotyrosine Rabbit anti-Human, Unconjugated, Clone: PY2870R, Abnova™

Catalog No. 89339732 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-732 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-732 Supplier Abnova Corporation Supplier No. RAB00693
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

Phosphotyrosine Monoclonal antibody specifically detects Phosphotyrosine in Human samples. It is validated for ELISA.

Specifications

Antigen Phosphotyrosine
Applications ELISA
Classification Monoclonal
Clone PY2870R
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human Phosphotyrosine
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.