Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phosphotyrosine, recombinant, Mouse, Alexa Fluor 647, Clone: 4G10,

Catalog No. 05777AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05777AF647 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05777AF647 Supplier MilliporeSigma™ Supplier No. 05777AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Phosphorylation plays an important role in regulating protein activities and various cellular signaling events. Posttranslational phosphorylation can occur on histidine (pHis), serine (pSer), threonine (pThr), and tyrosine (pTyr) residues. Due to the early advent of anti-pTyr antibodies, tyrosine phosphorylation remains the mostly studied signaling events in all biological research fields. Tyrosine phosphorylation is considered to be one of the key steps in signal transduction and regulation of enzymatic activity. Before the availability of anti-pTyr antibodies, protein tyrosine phosphorylation was routiniely detected by time-consuming radioactive labeling. Although radioactive labeling is still considered as more sensitive, advancement in detection technology has greatly improved the detection limits of antibody-based methods. Anti-pTyr antibodies are commonly used in ELISA, immunoprecipitation, immunoblotting (including dot blot and Western blot), as well as indirect immunofluourescent detection of tyrosine phosphorylation in tissue and cell samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphotyrosine, recombinant
Applications Immunocytochemistry
Classification Monoclonal
Clone 4G10
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 647
Formulation Purified mouse monoclonal IgG2a antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % Sodium Azide.
Host Species Mouse
Immunogen Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Purified via heavy chain C-terminus hexa-histidine tag by Nickel affinity matrices prior to conjugation with Alexa Fluor™ 647.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Recognizes tyrosine-phosphorylated proteins from all species.
Target Species All species
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.