Learn More
Sigma Aldrich Photoha(tm)-irg Methacrylated Hyaluronic Acid Bioink Kit With Irgacure
Catalog No. 9170791EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. 91-707-91EA
Supplier: Sigma Aldrich 9170791EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
PhotoHA(TM)-IRG bioink kit consists of methacrylated hyaluronic acid (HAMA) and Irgacure photoinitiator. Hyaluronic acid is the most abundant glycosaminoglycan in the body being an important component of several tissues throughout the body. While it is abundant in extracellular matrices, hyaluronic acid also contributes to tissue hydrodynamics, movement and proliferation of cells, and participates in a number of cell surface receptor interactions. The photoinitiator consists of Irgacure 2959 to be formulated in methanol (methanol not included), which allows UV crosslinking of the printed structure at 365 nm. PhotoHA(TM)-IRG provides native-like 3D HA gels, and the final gel stiffness can be customized by changing HA concentrations and crosslinking.PhotoHA is a trademark of Advanced BioMatrix, Inc.
Specifications
Quantity 1 Ea.
Synonym 3D Bioprinting; Bioink; HAMA; Hyaluronic acid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.