Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Photosynthesis and Respiration Chamber
Shop All Aldon Products

Click to view available options
:
Photosynthesis and Respiration Chamber
Description
Requires
NeuLog™ Oxygen Logger Sensor (S98243-8ND), CO2 Logger Sensor (S98243-3ND), and Light Logger Sensor (S98243-20ND); sensors not included
Supplied with respiration chamber, #0 and #7 rubber stoppers, instructions.
Specifications
Specifications
Product Type | Photosynthesis and Respiration Chamber |
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | Monitor the cycle of photosynthesis and respiration of plants using o2, co2 and light sensors over several days |
Includes | Rubber stoppers, instructions |
Quantity | 1 |
Dimensions (L x W x H) | 12 x 12 x 12 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction