Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHOX2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | PHOX2A |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PHOX2A Polyclonal antibody specifically detects PHOX2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PHOX2A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Growth and Development, Neuronal Cell Markers, Neuroscience | |
PBS (pH 7.2) and 40% Glycerol | |
401 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
aristaless (Drosophila) homeobox, aristaless homeobox (Drosophila), fibrosis ofextraocular muscles, congenital, 2, autosomal recessive, aristaless homeobox homolog, Aristaless homeobox protein homolog, arix homeodomain protein, ARIX1 homeodomain protein, ARIXNCAM2, CFEOM2MGC52227, paired mesoderm homeobox protein 2A, paired-like homeobox 2apaired-like (aristaless) homeobox 2a, PMX2AFEOM2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AAELLKAWQPAESGPGPFSGVLSSFHRKPGPALKTNLF | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title