Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHOX2B Mouse anti-Human, Mouse, Clone: 1F5F5, Proteintech
SDP

Catalog No. 501736687 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6687 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6687 Supplier Proteintech Group Inc Supplier No. 662541IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.

Specifications

Antigen PHOX2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F5F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHOX2B
Gene Accession No. O35690, Q99453
Gene Alias NBPhox, Neuroblastoma Phox, paired like homeobox 2b, PHOX2B, PHOX2B homeodomain protein, PMX2B
Gene Symbols Phox2b
Host Species Mouse
Immunogen PHOX2B Fusion Protein Ag13196
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18935, 8929
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.