Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHOX2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121236 Supplier Invitrogen™ Supplier No. PA5121236
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: PC-12, U-251MG. The target is usually found in the following locations: Nucleus. Immunogen sequence: MYKMEYSYLN SSAYESCMAG MDTSSLASAY ADFSSCSQAS GFQYNPIRTT FGATSGCPSL TPGSCSLGTL RDHQSSPYAA VPYKLFTDHG.

The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHOX2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHOX2B
Gene Accession No. O35690, Q99453
Gene Alias dilated pupils 1; Dilp1; GENA 269; LOW QUALITY PROTEIN: paired mesoderm homeobox protein 2B; NBLST2; NBPhox; neuroblastoma paired-type homeobox protein; neuroblastoma Phox; paired like homeobox 2B; paired mesoderm homeobox 2b; paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B; PHOX2B homeodomain protein; Pmx2b
Gene Symbols PHOX2B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human PHOX2B (NP_003915.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18935, 364152, 8929
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.