Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHOX2B Rabbit anti-Human, Mouse, Clone: 4W5J8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PHOX2B |
---|---|
Clone | 4W5J8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PHOX2B Monoclonal antibody specifically detects PHOX2B in Human, Mouse samples. It is validated for Western BlotSpecifications
PHOX2B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
8929 | |
IgG | |
Affinity purified |
4W5J8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein | |
A synthetic peptide corresponding to a sequence within amino acids 215-314 of human PHOX2B (Q99453). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title