Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHOX2B Recombinant Rabbit Monoclonal Antibody (4W5J8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535695
Description
Immunogen sequence: GGGPSPAGAP GAAGPGGPGG EPGKGGAAAA AAAAAAAAAA AAAAAAGGLA AAGGPGQGWA PGPGPITSIP DSLGGPFASV LSSLQRPNGA KAALVKSSMF.
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
Specifications
PHOX2B | |
Recombinant Monoclonal | |
2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35690, Q99453 | |
PHOX2B | |
A synthetic peptide corresponding to a sequence within amino acids 215-314 of human PHOX2B (NP_0039152). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
4W5J8 | |
Unconjugated | |
PHOX2B | |
dilated pupils 1; Dilp1; GENA 269; LOW QUALITY PROTEIN: paired mesoderm homeobox protein 2B; NBLST2; NBPhox; neuroblastoma paired-type homeobox protein; neuroblastoma Phox; paired like homeobox 2B; paired mesoderm homeobox 2b; paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B; PHOX2B homeodomain protein; Pmx2b | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18935, 8929 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction