Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHOX2B Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310477100UL
Description
PHOX2B Polyclonal specifically detects PHOX2B in Rat samples. It is validated for Western Blot.Specifications
PHOX2B | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat PHOX2B (XP_001077800). Peptide sequence MYKMEYSYLNSSAYESCMAGMDTSSLASAYADFSSCSQASGFQYNPIRTT | |
100 μg | |
Neuroscience | |
8929 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction