Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHPT1 Antibody (007), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB334596 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334596 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334596 Supplier Novus Biologicals Supplier No. NBP290109APC
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PHPT1 Monoclonal antibody specifically detects PHPT1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PHPT1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 007
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human PHPT1 (Uniprot#: Q9NRX4-1, Ala2-Tyr125)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 29085
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.