Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHPT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318414
This item is not returnable.
View return policy
Description
PHPT1 Polyclonal antibody specifically detects PHPT1 in Human, Mouse samples. It is validated for Western BlotSpecifications
PHPT1 | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1:10000 - 1:25000 | |
bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
Recognizes region between residues Between between 75 and 125.. | |
100 μg | |
Lipid and Metabolism | |
29085 | |
Store at 4°C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction