Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570708 Supplier Invitrogen™ Supplier No. PA570708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

PHR1 is a transcription factor known to participate in the regulation of genes involved in phosphate homeostasis and the regulation of genes involved in inter organ sulfate transport. Diseases associated with PHR1 protein mutation include Wallerian Degeneration and deafness due to Autosomal Recessive 63 dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Plekhb1
Gene Accession No. Q9UF11
Gene Alias AI315628; evectin-1; Evt1; evt-1; KPL1; MGC128483 protein; PH domain containing protein in retina 1; PH domain containing, retinal 1; PH domain-containing family B member 1; PH domain-containing protein in retina 1; Phr1; PHRET1; pleckstrin homology domain containing B1; pleckstrin homology domain containing, family B (evectins) member 1; pleckstrin homology domain retinal protein 1; Pleckstrin homology domain-containing family B member 1; Plekhb1; retinal 1
Gene Symbols Plekhb1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human PHR1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58473
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.