Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHR1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519042
Description
This antibody is predicted to react with bovine, human and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
PHR1 is a transcription factor known to participate in the regulation of genes involved in phosphate homeostasis and the regulation of genes involved in inter organ sulfate transport. Diseases associated with PHR1 protein mutation include Wallerian Degeneration and deafness due to Autosomal Recessive 63 dysfunction.
Specifications
PHR1 | |
Polyclonal | |
Unconjugated | |
Plekhb1 | |
AI315628; evectin-1; Evt1; evt-1; KPL1; MGC128483 protein; PH domain containing protein in retina 1; PH domain containing, retinal 1; PH domain-containing family B member 1; PH domain-containing protein in retina 1; Phr1; PHRET1; pleckstrin homology domain containing B1; pleckstrin homology domain containing, family B (evectins) member 1; pleckstrin homology domain retinal protein 1; Pleckstrin homology domain-containing family B member 1; Plekhb1; retinal 1 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
64471 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9WU68 | |
Plekhb1 | |
Synthetic peptide sequence (PPDSALESPFEE) corresponding to the N-terminus amino acids of PLEKHB1 (aa 8-19). | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction