Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5119698

This item is not returnable.
View return policy
Description
Positive Control: Rice tissue. Subcellular Location: Cytoplasm, Nucleus.
PHR2 Polyclonal antibody specifically detects PHR2 in Plant samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
PHR2 | |
Polyclonal | |
Unconjugated | |
LOC4332801 | |
Rabbit | |
Protein A | |
RUO | |
4332801 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
OsPHR2; protein phosphatase starvation response 2 | |
Recombinant protein within rice PHR2 aa 1-300 | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction