Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116730 Supplier Invitrogen™ Supplier No. PA5116730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Juakat, K-562, HepG2, HeLa Immunogen sequence: KAEAPSSPDV APAGKEDSPS ASGRVQEAAR PEEVVSQTPL LRSRALVKRV TWNLQESESS APAEDRAPRA PLHRPQKPRE GAWDMEDVAP TGVRQVFSEL P.

Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHRF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PHRF1
Gene Accession No. Q9P1Y6
Gene Alias AA673488; CTD-binding SR-like protein rA9; Kiaa1542; PHD and RING finger domain-containing protein 1; PHD and ring finger domains 1; phrf1; PPP1R125; protein phosphatase 1, regulatory subunit 125; RNF221
Gene Symbols PHRF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1350-1450 of human PHRF1 (NP_0012735121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.