Learn More
PHRF1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302044A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.Specifications
PHRF1 | |
Polyclonal | |
Unconjugated | |
PHRF1 | |
CTD binding SR like protein rA9, CTD-binding SR-like protein rA9, KIAA1542, PPP1R125, protein phosphatase 1, regulatory subunit 125, RNF221 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101471, 57661 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
A6H619, Q9P1Y6 | |
PHRF1 | |
Between 1400 and 1450 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.