Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ pHrodo™ AM Variety Pack

Catalog No. P35380
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P35380 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P35380 Supplier Invitrogen™ Supplier No. P35380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quantify intracellular pH with the pHrodo AM Variety Pack, which contains pHrodo Red and Green AM intracellular pH indicator dyes that permeate cells and fluoresce in acidic cytosolic environments. These pHrodo AM dyes are compatible with HCS, HTS, fluorescence microplate readers, and flow cytometry.

Measure and quantify intracellular pH with the pHrodo AM Variety Pack, which includes pHrodo Red and Green AM indicator dyes, fluorogenic probes that traverse the cell membrane and remain within the intracellular space upon cleavage by nonspecific esterases.

The pHrodo AM Variety Pack contains one vial each of the pHrodo Red AM and pHrodo Green AM indicator dyes, with each vial providing enough reagent volume to test one 96-well plate. pHrodo Red AM and pHrodo Green AM intracellular pH indicators are fluorogenic probes used to measure intracellular pH in live cells. They are weakly fluorescent at neutral pH, but dramatically increase in brightness as the pH decreases. These reagents can quantify cellular cytosolic pH in the range of 4–9 with a pKa of ∼6.5 and have excitation/emission spectra of 560/585 nm (pHrodo Red) and 509/533 nm (pHrodo Green).

pHrodo Red AM and pHrodo Green AM dyes are photostable and appropriate for use in multiplexing experiments; pHrodo Red easily multiplexes with a wide variety of blue, green, and far-red dyes and reporters such as GFP, Fuo-4, calcein, NucBlue, CellEvent Caspase 3/7 green, Mitosox Green, and Mitotracker Deep Red, among many others. pHrodo Green dye easily multiplexes with a wide variety of blue, red, and far-red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others. The pHrodo Red and pHrodo Green dyes have been modified with AM ester groups, which results in an uncharged molecule that can permeate cell membranes. Once inside the cell, the lipophilic blocking groups are cleaved by nonspecific esterases, resulting in a compound that is retained within the intracellular space. Fluorescence intensity of the probe is then an indicator of intracellular pH. Subsequent use of the Intracellular pH Calibration Buffer Kit (Cat. No. P35379) allows this intracellular pH to be quantified.

pHrodo Red AM and pHrodo Green AM intracellular pH indicators are compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), flow cytometry, and microplate-based fluorimetry or high throughput screening (HTS). These reagents are also compatible with widefield or confocal fluorescence microscopes, flow cytometers, fluorescent plate readers, and high content instruments.

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-permeant
Color Green, Orange
Content And Storage Store at 2–8°C, desiccate, and protect from light.

Description pHrodo™ AM Variety Pack
Detection Method Fluorescence
For Use With (Application) Cell Analysis
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
Product Type Fluorogenic Probe
Form Liquid
Format 96-well Plate
Indicator pH Indicator
Indicator Properties Fluorescent Dye-Based
Product Line pHrodo
Quantity 1 Kit
Solubility DMSO (Dimethylsulfoxide)
Sub Cellular Localization Cytoplasm
Sufficient For Testing one 96-well plate
Excitation/Emission pHrodo™ Green: 509/533 nm, pHrodo™ Red: 560/585 nm
Emission Wavelength Visible
Show More Show Less

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.