Learn More
Invitrogen™ pHrodo™ Red Phagocytosis Particle Labeling Kit for Flow Cytometry
Description
- Specifically detect phagocytosis and endocytosis with pH-sensitive fluorogenic dye - discriminate endcytosed from adherent and extracellular particles
- Reduced signal variability and improved timing in sensitive experiments - no need for wash steps or quencher dye
- Bright red fluorescence - conveniently multiplex with green dyes such as GFP, Fluo-4, or calcein
The new Molecular Probes™ proprietary pH-sensitive rhodamine-based pHrodo™ Red dye is non-fluorescent at neutral pH, but turns bright red upon acidification. Because it is both fluorogenic and pH-sensitive, the pHrodo™ Red dye can be used as a specific sensor of phagocytic events; acidification of the phagosome following phagocytosis is marked by red fluorescence. It is therefore an ideal tool with which to study phagocytosis and its regulation by drugs and/or environmental factors.
Rapid and reproducible
Wash steps and quencher dyes are not needed since the pHrodo™ Red dye is non-fluorescent outside the cell, making the staining protocol simple and fast. The elimination of wash steps and quencher dyes also improves assay reproducibility, particularly in plate reader-based assays.
Flexible applications
Use ready-made pHrodo™ Red BioParticles™ conjugates for convenient analysis of phagocytosis of Gram-positive or Gram-negative bacteria, or the amine-reactive pHrodo™ Red SE form for labeling microorganisms or proteins of your choice.
Compatible with multiple platforms
pHrodo™ Red dye conjugates can be used in plate readers, fluorescence microscopy imaging, and flow cytometry applications. The pHrodo™ Red Phagocytosis Particle Labeling Kit and the pHrodo™ Red E. coli BioParticles™ Phagocytosis Kit for flow cytometry were designed for rapid and convenient measurements of phagocytic activity in whole blood samples by flow cytometry. The kits include all the reagents required for assessing particle ingestion and red blood cell lysis. The Labeling Kit also contains the reagents required for labeling microorganisms with the pHrodo™ Red dye. Sufficient reagents are provided in both kits for approximately 100 assays.
The optimal absorption and fluorescence emission maxima of the pHrodo™ Red BioParticles™ conjugate is approximately 560nm and 585nm, respectively. However, the fluorophore is readily excited with the 488nm argon-ion laser installed on most flow cytometers.
Specifications
Specifications
| Content And Storage | Contains lysis buffer (10 mL), buffer B (200 mL), wash buffer (200 mL), 1 vial pHrodo™ red succinimidyl ester (1 mg, lyophilized product), 1 vial DMSO (0.5 mL), 1 bottle sodium bicarbonate (50 mL, 0.1 M, pH 9.3). Store in refrigerator |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Red Phagocytosis Particle Labeling Kit |
| Dye Type | pHrodo™ Red |
| Format | Bottle(s) |
| Product Line | pHrodo |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |
| Emission Wavelength | 560/585 |
Frequently Asked Questions (FAQs)
pHrodo is relatively non-fluorescent until it enters the acidic phagosome, at which point its fluorescence increases. If you fix the sample, the pHrodo will only reflect the pH of the buffer the cells are in, and not the pH of the phagosome. For this reason, we do not recommend fixing samples. If you want to see how many cells engulfed the labeled bacteria, fix the cells and then place the fixed cells in an acidic buffer for the assay.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.