Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pht1 / Histone H2A.Zac (pan-acetyl) Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501992755
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2755 100 μL
50-199-2756 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2755 Supplier Active Motif Supplier No. 39642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.

Specifications

Antigen Pht1 / Histone H2A.Zac (pan-acetyl)
Applications Dot Blot, Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
Gene HTZ1
Gene Accession No. Q12692
Gene Symbols HTZ1
Host Species Rabbit
Immunogen This Pht1 / H2AZ acetyl antibody was raised against a peptide containing acetyl-lysine at positions 5, 7, 12 and 16 from the amino terminus of Pht1, the histone H2AZ variant in S. pombe.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 854150
Target Species Yeast
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.