Learn More
PHTF1 Monoclonal Antibody (3B7)

Description
PHTF1 Monoclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | PHTF1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | PHTF1 |
| Gene Accession No. | Q9UMS5 |
| Gene Alias | PHTF; PHTF1; Putative homeodomain transcription factor 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.