Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalamidoxime, 97%, Thermo Scientific™

Catalog No. AAH6634203
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885960-32-7
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.194
InChI Key CQHMIKPVOMZRKW-KQQUZDAGSA-N
Synonym 1,2-bis carboxamidoximo benzene
PubChem CID 6346926
IUPAC Name N-[(E)-amino-[(6E)-6-[amino(nitroso)methylidene]cyclohexa-2,4-dien-1-ylidene]methyl]hydroxylamine
SMILES C1=CC(=C(N)NO)C(=C(N)N=O)C=C1

Specifications

Color Cream
Quantity 1 g
Formula Weight 194.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Phthalamidoxime
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.