Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalan, 96%, Thermo Scientific™

Catalog No. AA4559214
Change view
Click to view available options
Quantity:
25g
Catalog No. Quantity
AA4559214 25g
1 options

Catalog No. AA4559214

Supplier: Thermo Scientific Chemicals 4559214

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 496-14-0
Density 1.094 g/mL
Boiling Point 192°C
Flash Point 63°C (145°F)
Molecular Formula C8H8O
Refractive Index 1.546
Quantity 25g
MDL Number MFCD00005932
Beilstein 111932
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
SMILES C1C2=CC=CC=C2CO1
IUPAC Name 1,3-dihydro-2-benzofuran
Molecular Weight (g/mol) 120.151
PubChem CID 10327
Formula Weight 120.15
Percent Purity 96%
Chemical Name or Material Phthalan
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 207-815-2
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.