Learn More
Phthalan, 98%, Thermo Scientific Chemicals
Description
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butylbiphenyl, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butylbiphenyl, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.
Solubility
Low aqueous solubility.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Density | 1.094 |
Boiling Point | 192°C |
Flash Point | 63°C (145°F) |
Refractive Index | 1.546 |
Quantity | 5 g |
Beilstein | 111932 |
Solubility Information | Low aqueous solubility. |
Formula Weight | 120.15 |
Percent Purity | 98% |
Chemical Name or Material | Phthalan |
Safety and Handling
Hazard Category | H227 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P280-P370+P378q-P501c |
EINECSNumber | 207-815-2 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.