Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalate Esters Mix, SPEX CertiPrep™
SDP

Catalog No. 8061x Shop All Spex CertiPrep Products
Click to view available options
Quantity:
1 mL

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

AnalyteCAS No.Concentration
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Bis(2-methoxyethyl)phthalate117-82-82000μg/mL
Bis(2-n-butoxyethyl)phthalate117-83-92000μg/mL
Bis(4-methyl-2-pentyl)phthalate146-50-92000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Diamyl phthalate131-18-0
Chemical Name or Material Phthalate Esters Mix
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max % 97.0, 0.2, 0.2, 0.2, 0.2, 0.2
CAS 84-61-7, 117-83-9, 84-76-4, 146-50-9, 84-75-3
Methods For US EPA Methods 625, 8270C
Analyte or Component Names bis(2-Ethylhexyl)phthalate,Bis(2-methoxyethyl )phthalate,Bis(2-n-butoxyethyl)phthalate
Technique AA, ICP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Element bis(2-Ethylhexyl)phthalate, bis(2-methoxyethyl )phthalate, bis(2-n-butoxyethyl)phthalate
Solvent or Matrix Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.