Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Phthalic acid mono-2-ethylhexyl ester

Catalog No. 796832500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-683-2500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-683-2500MG Supplier Sigma Aldrich Supplier No. 796832500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Phthalic acid mono-2-ethylhexyl ester

Specifications

CAS 4376-20-9
Density 1.0864 g/mL (at 25°C)
Molecular Formula C16H22O4
Refractive Index n/D 1.5051
Linear Formula C16H22O4
MDL Number MFCD00041500
Quantity 100 mg
Synonym mono-2-Ethylhexyl phthalate
Molecular Weight (g/mol) 278.34
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.