Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimide, 99%

Catalog No. p-3593491
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 85-41-6 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-41-6
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005881
InChI Key XKJCHHZQLQNZHY-UHFFFAOYSA-N
Synonym phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
PubChem CID 6809
ChEBI CHEBI:38817
IUPAC Name isoindole-1,3-dione
SMILES O=C1NC(=O)C2=CC=CC=C12

Specifications

Melting Point >233.5°C
Color White to Yellow
Boiling Point 366.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 21, 458
Fieser 07,330
Merck Index 15, 7485
Sublimation Point 0366.0°C
Solubility Information Solubility in water: 0.4-0.6g/L (20°C). Other solubilities: 5 g/100g boiling alcohol, freely soluble in aqueous alkali hydroxides, soluble in boiling acetic acid, acetone, hot ether, and dimethyl sulfoxide, almost insoluble in benzene, petroleum ether
Formula Weight 147.13
Percent Purity 99%
Physical Form Crystalline Flakes
Chemical Name or Material Phthalimide
Show More Show Less
EINECSNumber 201-603-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.