Learn More
Thermo Scientific™ Phusion™ Plus DNA Polymerase PROMO
Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing.
Supplier: Thermo Scientific™ F630L
Description
Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus DNA Polymerase to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance.
- High fidelity–provides >100X higher sequence accuracy than Taq polymerase
- Convenient setup–enables a universal annealing temperature of 60°C in PCR
- Enhanced specificity–hot-start mechanism reduces non-specific amplification and primer degradation
- Benchtop stability–automation enabled due to 24-hour stability of assembled reactions
- High inhibitor tolerance–works with DNA of suboptimal purity
The novel Phusion Plus reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus DNA Polymerase and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.
Phusion Plus DNA Polymerase is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.
Applications
- High-fidelity PCR
- Cloning
- Template generation for sequencing
- GC-rich PCR
- Long-range PCR (up to 20 kb)
- Mutagenesis
- High-throughput PCR
To simplify PCR reaction setup, Phusion Plus DNA Polymerase is also available in ready-to-use master mix formats. Phusion Plus PCR Master Mix (Cat. No. F631S) is a convenient 2X mix of enzyme, reaction buffer, magnesium chloride, and dNTPs. Only template and primers need to be added to the PCR reaction. Phusion Plus Green PCR Master Mix (F632S) additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.
Specifications
Phusion Plus DNA Polymerase (250 μL), 5X Phusion Plus Buffer (5 x 1.25 mL), 5X Phusion GC Enhancer (4 x 1.25 mL), Store at -15°C to -25°C. | |
Phusion Plus DNA Polymerase | |
Dry Ice | |
>100X | |
500 Reactions | |
Separate components |
High | |
15 to 30 sec./kb | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
Blunt | |
20 kb or less |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.