Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion™ Plus PCR Master Mix PROMO

Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs.

Specifications

Concentration 2X
GC-Rich PCR Performance High
Polymerase Phusion Plus DNA Polymerase
Reaction Speed 15 to 30 sec./kb
Shipping Condition Dry Ice
View More Specs

Products 2
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Price Quantity & Availability  
F631LPM
View Documents
Thermo Scientific™
F631L
500 Reactions N/A N/A N/A
F631SPM
View Documents
Thermo Scientific™
F631S
100 Reactions N/A N/A N/A
Description

Description

Thermo Scientific Phusion Plus PCR Master Mix is a convenient, ready-to-use 2X mixture of Phusion Plus DNA Polymerase, buffer, magnesium chloride, and dNTPs. The master mix format simplifies reaction setup with minimal pipetting steps and retains all features of the stand-alone format.

  • High fidelity–provides >100X greater sequence accuracy than Taq polymerase
  • Convenient setup–enables a universal annealing temperature of 60°C in PCR
  • Enhanced specificity–hot-start modification reduces non-specific amplification and primer degradation
  • Benchtop stability–automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance–works with DNA of suboptimal purity

Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus PCR Master Mix to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance. Its novel reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus PCR Master Mix and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus PCR Master Mix is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications

  • High-fidelity PCR
  • Cloning
  • Template generation for sequencing
  • GC-rich PCR
  • Long-range PCR (up to 20 kb)
  • Mutagenesis
  • High-throughput PCR
Specifications

Specifications

2X
Phusion Plus DNA Polymerase
Dry Ice
>100X
Blunt
20 kb or less
High
15 to 30 sec./kb
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Ready to use master mix
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.