Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion Site-Directed Mutagenesis Kit

Catalog No. F541
Change view
Click to view available options
Quantity:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F541 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F541 Supplier Thermo Scientific™ Supplier No. F541
Add to Cart
Edge
Add to Cart

Generate mutations with this fast and reliable system.

Phusion Site-Directed Mutagenesis Kit is a versatile and efficient tool for introducing point mutations, insertions, or deletions in any type of plasmid DNA. With this kit, the entire plasmid is amplified using phosphorylated primers that introduce the desired changes. The amplified, linear PCR product, containing the desired mutation, is circularized in a 5-minute ligation reaction with T4 DNA Ligase. The resulting plasmid can be then transformed into any competent E. coli cells.

Highlights

  • Robust and reliable exponential amplification method
  • No requirements, such as special vectors, restriction sites, or methylation status for the target plasmid
  • No need to destroy the starting template in a separate step
  • Phusion Hot Start II High Fidelity DNA Polymerase minimizes unwanted secondary mutations
  • Amplification of large plasmids up to 10 kb
  • Hot start modification of the polymerase prevents amplification of non-specific products and unwanted degradation of primers prior to first cycle of PCR
  • T4 DNA Ligase included in the kit; no purification steps before or after ligation
  • Compatible with all strains of competent E. coli cells
TRUSTED_SUSTAINABILITY

Specifications

Form Solution
Format Kit
GC-Rich PCR Performance High
Polymerase Phusion Hot Start II High Fidelity DNA Polymerase
Reaction Speed Fast
Mutagenesis Capability Single-Site Mutagenesis
Product Line Phusion
Product Type Site-Directed Mutagenesis Kit
Quantity 20 reactions
Fidelity (vs. Taq) 52X
Hot Start Yes
No. of Reactions 20 Reactions
Starting Material DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.