Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phycoerythrin Polyclonal Antibody, FITC
GREENER_CHOICE

Catalog No. PIPA535007
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535007 Supplier Invitrogen™ Supplier No. PA535007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-35007 targets Phycoerythrin-R/R-PE in FACS and IHC applications and shows reactivity with Tag samples. The PA5-35007 immunogen is highly purified PE.

Anti-RPE (R-Phycoerythrin) antibodies can be used to bind conjugated primary antibody without decreasing fluorescent intensity of the RPE fluorochrome. This method is used for separation of cells labeled with RPE-conjugated antibodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phycoerythrin
Applications Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene Alias PE; RPE; R-PE; R-phycoerythrin
Host Species Rabbit
Immunogen highly purified PE.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.