Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHYH Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300120AF700
Description
PHYH Polyclonal specifically detects PHYH in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PHYH | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
LNAP1, PAHXLN1, peroxisomal, PhyH, PHYH1, phytanoyl-CoA 2-hydroxylase, phytanoyl-CoA hydroxylase, phytanoyl-CoA hydroxylase (Refsum disease), RD | |
Produced in rabbits immunized with purified, recombinant PHYH (O14832; Ser31-Leu338). | |
0.1 mL | |
Lipid and Metabolism, Vision | |
5264 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction