Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHYH Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA530748
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530748 Supplier Invitrogen™ Supplier No. MA530748
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PHYH.

PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHYH
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Phyh
Gene Accession No. O14832
Gene Alias 1600020H07Rik; 2-HPCL; AI256161; AI265699; HPCL; HPCL2; induced by chronic shear stress; LN1; Lnap1; LOC100056651; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; Pahx; PHYH; PHYH protein; PHYH1; PHYH2; phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; phytanoyl-CoA 2-hydroxylase; phytanoyl-CoA alpha-hydroxylase; phytanoyl-CoA dioxygenase peroxisomal; phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase; phytanoyl-CoA hydroxylase (Refsum disease); RD; zgc:110203
Gene Symbols Phyh
Host Species Mouse
Immunogen Recombinant Human PHYH protein (Ser31-Leu338).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5264
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.