Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHYH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582306
Description
Immunogen sequence: LPGTHKGSLK PHDYPKWEGG VNKMFHGIQD YEENKARVHL VMEKGDTVFF HPLLIHGSGQ NKTQGFRKAI SCHFASADCH YIDVKGTSQE NIEKEVVGIA HKFFGAENSV NLKDIWMFRA.
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
Specifications
PHYH | |
Polyclonal | |
Unconjugated | |
Phyh | |
1600020H07Rik; 2-HPCL; AI256161; AI265699; HPCL; HPCL2; induced by chronic shear stress; LN1; Lnap1; LOC100056651; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; Pahx; PHYH; PHYH protein; PHYH1; PHYH2; phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; phytanoyl-CoA 2-hydroxylase; phytanoyl-CoA alpha-hydroxylase; phytanoyl-CoA dioxygenase peroxisomal; phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase; phytanoyl-CoA hydroxylase (Refsum disease); RD; zgc:110203 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16922, 5264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14832, O35386 | |
Phyh | |
Recombinant protein corresponding to Human PHYH. Recombinant protein control fragment (Product #RP-89687). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction