Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PHYH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582436
Description
Immunogen sequence: TSGTISSASF HPQQFQYTLD NNVLTLEQRK FYEENGFLVI KNLVPDADIQ RFRNEFEKIC RKEVKPLGLT VMRDVTISKS EYAPSEKMIT KVQDFQEDKE LFRYCTLPEI LKYVECFTGP NIMAMHTMLI NKPPDS.
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
Specifications
PHYH | |
Polyclonal | |
Unconjugated | |
Phyh | |
1600020H07Rik; 2-HPCL; AI256161; AI265699; HPCL; HPCL2; induced by chronic shear stress; LN1; Lnap1; LOC100056651; Lupus nephritis-associated peptide 1; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; Pahx; PHYH; PHYH protein; PHYH1; PHYH2; phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; phytanoyl-CoA 2-hydroxylase; phytanoyl-CoA alpha-hydroxylase; phytanoyl-CoA dioxygenase peroxisomal; phytanoyl-CoA dioxygenase, peroxisomal; phytanoyl-CoA hydroxylase; phytanoyl-CoA hydroxylase (Refsum disease); RD; zgc:110203 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14832 | |
Phyh | |
Recombinant protein corresponding to Human PHYH. Recombinant protein control fragment (Product #RP-89686). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction