Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHYHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121077 Supplier Invitrogen™ Supplier No. PA5121077
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse spleen, Mouse liver, Mouse kidney, Mouse heart, Mouse skin, Rat liver, Rat kidney. Immunogen sequence: MACLSPSQLQ KFQQDGFLVL EGFLSAEECV AMQQRIGEIV AEMDVPLHCR TEFSTQEEEQ LRAQGSTDYF LSSGDKIRFF FEKGVFDEKG NFLVPPEKSI NKIGHALHAH DPVFKSITHS FKVQTLARSL GLQMPVVVQS MYIFKQPHFG GEVSPHQDAS FLYTEPLGRV LGVWIAVEDA TLENGCLWFI PGSHTSGVSR RMVRAPVGSA PGTSFLGSEP ARDNSLFVPT PVQRGALVLI HGEVVHKSKQ NLSDRSRQAY TFHLMEASGT TWSPENWLQP TAELPFPQLY T.

PHYHD1 (phytanoyl-CoA dioxygenase domain containing 1) is a 291 amino acid protein belonging to the PHYH family and the PHYHD1 subfamily. Encoded by a gene that maps to human chromosome 9q34.11, PHYHD1 exists as three alternatively spliced isoforms and likely functions as an alpha-ketoglutatare-dependent dioxygenase. PHYHD1 participates in metal ion binding and oxidoreductase activity, thereby acting on single donors with incorporation of two atoms of oxygen. Related to PHYH (phytanoyl-CoA 2-hydroxylase), an Fe(II) and 2-oxoglutarate (2OG)-dependent oxygenase that catalyzes the initial alpha-oxidation step in the degradation of phytenic acid in peroxisomes, PHYHD1 similarly exhibits additional homologues in a wide range of metazoans and bacteria. PHYHD1 also may play a role in DNA methylation in early postnatal liver development and mammalian differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHYHD1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PHYHD1
Gene Accession No. Q5BJP9, Q5SRE7, Q9DB26
Gene Alias Phyhd1; phytanoyl-CoA dioxygenase domain containing 1; phytanoyl-CoA dioxygenase domain-containing protein 1
Gene Symbols PHYHD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-291 of human PHYHD1 (NP_001094346.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227696, 254295, 296621
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.