Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHYHIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531835
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531835 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531835 Supplier Invitrogen™ Supplier No. PA531835
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, PC-3, SK-N-SH, mouse brain. Predicted reactivity: Human (98%), Mouse (98%), Rat (98%), Rhesus Monkey (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Its interaction with PHYH suggests a role in the development of the central system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHYHIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PHYHIP
Gene Accession No. Q8K0S0, Q92561
Gene Alias AW049870; C630010D02Rik; DYRK1A interacting protein 3; DYRK1AP3; KIAA0273; Lnap1ip; lupus nephritis-associated peptide 1 interacting protein; PAHX-AP; PAHX-AP#1; PAHXAP1; PAHX-AP1; PHYHIP; phytanoyl-CoA 2-hydroxylase interacting protein; phytanoyl-CoA alpha-hydroxylase associated protein; phytanoyl-CoA hydroxylase interacting protein; phytanoyl-CoA hydroxylase-associated protein 1; Phytanoyl-CoA hydroxylase-interacting protein
Gene Symbols PHYHIP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PHYHIP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 105653, 9796
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.