Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PHYHIP Polyclonal Antibody

Supplier: Thermo Scientific PA5113584
Description
Specifications
| PHYHIP | |
| Polyclonal | |
| Unconjugated | |
| PHYHIP | |
| AW049870; C630010D02Rik; DYRK1A interacting protein 3; DYRK1AP3; KIAA0273; Lnap1ip; lupus nephritis-associated peptide 1 interacting protein; PAHX-AP; PAHX-AP#1; PAHXAP1; PAHX-AP1; PHYHIP; phytanoyl-CoA 2-hydroxylase interacting protein; phytanoyl-CoA alpha-hydroxylase associated protein; phytanoyl-CoA hydroxylase interacting protein; phytanoyl-CoA hydroxylase-associated protein 1; Phytanoyl-CoA hydroxylase-interacting protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q92561 | |
| PHYHIP | |
| Synthetic peptide directed towards the N terminal region of human PHYHIP (aa 51-100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction