Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHYHIPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570458
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570458 Supplier Invitrogen™ Supplier No. PA570458
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 100%.

PHYHIPL may play a role in the development of the central system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHYHIPL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PHYHIPL
Gene Accession No. Q96FC7
Gene Alias 4921522K17Rik; AI267048; KIAA1796; PHYHIPL; phyhiplb; phytanoyl-CoA 2-hydroxylase interacting protein like; phytanoyl-CoA 2-hydroxylase interacting protein-like; phytanoyl-CoA 2-hydroxylase interacting protein-like b; phytanoyl-CoA hydroxylase interacting protein-like; Phytanoyl-CoA hydroxylase-interacting protein-like; Unknown (protein for MGC:134205); UNQ6309/PRO20934; zgc:100983
Gene Symbols PHYHIPL
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human PHYHIPL.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84457
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.