Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PHYKPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584606 Supplier Invitrogen™ Supplier No. PA584606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLNVLEKEQL QDHATSVGSF LMQLLGQQKI KHPIVGDVRG V Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 73%.

This is a nuclear gene encoding a mitochondrial enzyme that catalyzes the conversion of 5-phosphonooxy-L-lysine to ammonia, inorganic phosphate, and 2-aminoadipate semialdehyde. Mutations in this gene may cause phosphohydroxylysinuria. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHYKPL
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHYKPL
Gene Accession No. Q8IUZ5
Gene Alias 2900006B13Rik; 5-phosphohydroxy-L-lysine phospholyase; 5-phosphohydroxy-L-lysine phospho-lyase; 5-phosphonooxy-L-lysine phospho-lyase; Agxt2l2; Alanine--glyoxylate aminotransferase 2-like 2; alanine-glyoxylate aminotransferase 2-like 2; PHLU; PHYKPL; PP9286
Gene Symbols PHYKPL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PHYKPL. Recombinant protein control fragment (Product #RP-107262).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85007
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.